Learn More
Macherey-Nagel™ Visocolor™ Eco Hardness (Total) Test Kit
Provides titrimetric determination of total hardness. Macherey Nagel™ Visocolor™ Eco Hardness (Total) Test Kits are packed in an environmentally friendly box and contain easy to understand instructions in 6 languages.
Brand:  Macherey-Nagel™ 931029
Additional Details : CAS Number : 1034-39-5 Weight : 1.00000kg Transport : UN number : 3316 Chem class : 9 Pack group : II
Product Code. 10694701
  • 580.00 SEK / Each
Estimated Shipment: 05-06-2024
to see stock.
Add to basket
Description
Description
  • In acc. with DIN 38406 E3 and DIN 38409 H6
  • Allow enough water to run through copper pipes prior to sampling
  • Reagent bottles with clear dosing instructions
Specifications
Specifications
The total hardness of water is based on its content of alkaline earth ions (calcium and magnesium ions)
1 drop = 1 °d
ECO Hardness (total) Titrimetric Test Kit
1 Each
110 Tests with an average hardness of 10 °d
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • VISOCOLOR ECO Total Hardness
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
  • MIXTURE LIST-Contains : ethanol