Learn More
Macherey-Nagel™ Visocolor™ ECO Nitrate Colorimetric Test Kit
Refill kit for 110 determinations over the measuring range 1 to 120mg/L NO3
Brand:  Macherey-Nagel™ 931241
Additional Details : Weight : 0.50000kg
Product Code. 10531943
  • 297.03 SEK / Each
Estimated Shipment: 15-05-2024
to see stock.
Specifications
Colorimetric Determination of Nitrate
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • VISOCOLOUR ECO nitrate, refill pack
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H411-Toxic to aquatic life with long lasting effects.
Supplemental information
  • MIXTURE LIST-Contains : zinc powder (stabilized)