Learn More
Macherey-Nagel™ Visocolor™ ECO Reagent Case
Prepacked with test kits for ammonium, carbonate and total hardness, nitrate, nitrite, pH and phosphate measurement. Macherey-Nagel™ Visocolor™ ECO Reagent Case is a rugged case with a premium foam inlay for storing and transporting reagents needed for water analysis.
Brand:  Macherey-Nagel™ 931301
Additional Details : Weight : 0.34200kg Transport : UN number : 3316 Chem class : 9 Pack group : II
Product Code. 11370585
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
Visicolor™ ECO Reagent Cases are flexible tools for easy and convenient handling of all areas of water and soil analysis. They enable users to get results fast and directly at the point of interest when testing the areas of environmental management and water monitoring.
  • Easy to handle mini-lab
  • For exact and accurate visual determination
  • Expandable with one additional Visocolor™ ECO test
  • Compact case for extensive analysis
  • No previous chemical knowledge/experience required
  • Case includes all necessary test instructions and analytical accessories
  • Photometer not included
Specifications
Specifications
Blue with Black foam
Visocolor™ ECO colorimetric test kits
ECO Test Kits: Ammonium, Carbonate Hardness, Total Hardness, Nitrate, Nitrite, pH, Phosphate
Various
Without photometer
Water analysis
1 Kit
Prepacked Reagent Case