missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Visocolor™ ECO Test Kit

Allows determination of even low limiting values with accuracy. Macherey-Nagel™ Visocolor™ ECO Test Kits is a group of colorimetric and titrimetric test kits that use precisely dosed single reagents. The results are evaluated visually with high-quality color comparison cards.

Brand:  Macherey-Nagel™ 931040

Additional Details : Weight : 0.45000kg Transport : UN number : 3316 Chem class : 9 Pack group : II


 View more versions of this product

Product Code. 10457292

  • 1080.00 SEK / 1 test
Estimated Shipment: 03-06-2024
to see stock.
Add to basket

Description

Description

Visocolor™ ECO colorimetric and titrimetric test kits achieve high sensitivity and accuracy by using single reagents that can be dosed precisely and by the ability to compensate for the turbidity and color of water samples. The results are evaluated visually with color comparison cards adjusted to the original colors of standard solutions or photometrically with the PF-3 and PF-12Plus photometers for quantitative evaluation.

Easy

  • Chemical analysis without further accessories
  • No extensive training necessary
  • Color-coded reagents with clear dosing instructions
  • Easy to understand instruction manual

Safe

  • Pictogram test instructions
  • Reaction basis according to international standards
  • Compensation of turbidity and color

Unique

  • High quality test kits
  • Business-prized refill packs available
  • Ecologically friendly disposal of used reagents
  • Delivered in cardboard box with plastic inlay
Specifications

Specifications

Dimethylglyoxime
PF-12 Plus
150 Determinations
0 to 1.5mg/L Ni2+ (Visual), 0.04 to 5.00mg/L Ni2+ (Photometric)
Colorimetric
Nickel determination
Reagents, Color comparator, Measuring spoon/glass, Syringe, Instructions in a cardboard box with plastic insert
1 Each
150 Determinations
ECO Nickel Colorimetric Test Kit
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • VISOCOLOUR ECO nickel

Signal Word
  • Danger

Hazard Category
  • Oxidising solid Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 A
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Specific target organ toxicity Category 3

Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : ammoniumm peroxydisulphate, sodium hydroxide

SDS
Documents

Documents

Certificates
Special Offers

Special Offers