missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Visocolor™ ECO Test Kit

Allows determination of even low limiting values with accuracy. Macherey-Nagel™ Visocolor™ ECO Test Kits is a group of colorimetric and titrimetric test kits that use precisely dosed single reagents. The results are evaluated visually with high-quality color comparison cards.

Brand:  Macherey-Nagel™ 931024

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10570403

  • 1178.64 SEK / 1 test
Estimated Shipment: 14-06-2024
to see stock.
Add to basket

Description

Description

Visocolor™ ECO colorimetric and titrimetric test kits achieve high sensitivity and accuracy by using single reagents that can be dosed precisely and by the ability to compensate for the turbidity and color of water samples. The results are evaluated visually with color comparison cards adjusted to the original colors of standard solutions or photometrically with the PF-3 and PF-12Plus photometers for quantitative evaluation.

Easy

  • Chemical analysis without further accessories
  • No extensive training necessary
  • Color-coded reagents with clear dosing instructions
  • Easy to understand instruction manual

Safe

  • Pictogram test instructions
  • Reaction basis according to international standards
  • Compensation of turbidity and color

Unique

  • High quality test kits
  • Business-prized refill packs available
  • Ecologically friendly disposal of used reagents
  • Delivered in cardboard box with plastic inlay
Specifications

Specifications

DEHA determination
Reagents, Color comparator, Measuring spoon/glass, Syringe, Instructions in a cardboard box with plastic insert
1 Each
Colorimetric
N/A
125 Determinations
0 to 0.30mg/L DEHA
ECO DEHA Colorimetric Test Kit
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers