missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Visocolor™ HE Colorimetric Test Kit

Provides the highest sensitivity and accuracy in visual analytics. Macherey-Nagel™ Visocolor™ HE Colorimetric Test Kits determine even the lowest limiting values using longer measuring tubes and larger sample volumes. Includes all reagents and testing equipment/supplies.

Brand:  Macherey-Nagel™ 920055

Additional Details : Weight : 0.40000kg Transport : UN number : 3316 Chem class : 9 Pack group : II


 View more versions of this product

Product Code. 10628061

  • 1315.00 SEK / 100 tests
Estimated Shipment: 13-05-2024
to see stock.
Add to basket

Description

Description

Visocolor™ HE test kits provide highly precise analysis via exact dosing of the single reagents as well as compensating for turbidity and color. The sensitivity of Visocolor™ HE is 10 to 100 times higher compared to other Visocolor tests.

  • Visual evaluation
  • Environment-friendly, without toxic reagents
  • Economically priced
  • Convenient handling
  • Highest accuracy due to extremely narrow gradation
  • Highest sensitivity down to 0.002mg/L due to longer measuring tubes
  • Reagent bottles with clear dosing instructions
  • Compensation of turbidities and colors
  • Refill packs available
Specifications

Specifications

Manganese (Mn)
Highly sensitive
Manganese determination
Reagents and test equipment (e.g. glass tubes with screw caps, comparator block, color comparison disk)
1 Each
Formaldoxime
Water analysis test kit
Color change: White to Brown
HE Colorimetric Test Kit
0.0 - 0.03 - 0.06 - 0.10 - 0.15 - 0.20 - 0.25 - 0.30 - 0.40 - 0.50mg/L Mn
100
0 to 0.50mg/L
7 min.
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • VISOCOLOR HE Manganese

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H351-Suspected of causing cancer.
  • H371-May cause damage to organs.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : performaldehyde, hydroxylammonium chloride, ammonia solution, methanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers