missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VISTA Monoclonal Antibody (B7H5DS8), PE-eFluor™ 610, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
2020.00 SEK - 4980.00 SEK
Specifications
Antigen | VISTA |
---|---|
Clone | B7H5DS8 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16606464
|
Affymetrix eBioscience
61-1088-42 |
100 Tests |
4980.00 SEK
100 tests |
Estimated Shipment: 27-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16696454
|
Affymetrix eBioscience
61-1088-41 |
25 Tests |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
VISTA (V-domain Ig containing suppressor of T-cell activation), also known as B7-H5 or PD-1H, is a product of the VSIR gene. Although VISTA bears sequence homology to PD-L1 it has a different expression pattern. In both mouse and humans, VISTA is expressed predominantly on hematopoietic cells with the highest densities on myeloid cells, and lower levels on T cells. It is usually absent on B cells. VISTA expressed by antigen presenting cells has been shown to directly suppress proliferation and cytokine production of CD4 and CD8 T cells, making it a target for cancer immunotherapy. The reported ligands of VISTA include CD28H, VSIG-3 and VSIG-8.Specifications
VISTA | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
64115 | |
VISTA-Fc protein | |
Primary | |
Antibody |
B7H5DS8 | |
Flow Cytometry | |
PE-eFluor 610 | |
Mouse | |
Human | |
Q9H7M9 | |
VSIR | |
IgG1 κ | |
Affinity chromatography | |
VSIR |