missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ VISTA Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552493
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Immunogen sequence: LTFQDLHLHH GGHQAANTSH DLAQRHGLES ASDHHGNFSI TMRNLTLLDS GLYCCLVVEI RHHHSEHRVH GAMELQVQTG KDAPSNCVVY PSSS Highest antigen sequence identity to the following orthologs: Mouse - 60%, Rat - 59%.
VISTA (V-domain Ig containing suppressor of T-cell activation), also known as B7-H5 or PD-1H, is a product of the VSIR gene. Although VISTA bears sequence homology to PD-L1 it has a different expression pattern. In both mouse and humans, VISTA is expressed predominantly on hematopoietic cells with the highest densities on myeloid cells, and lower levels on T cells. It is usually absent on B cells. VISTA expressed by antigen presenting cells has been shown to directly suppress proliferation and cytokine production of CD4 and CD8 T cells, making it a target for cancer immunotherapy. The reported ligands of VISTA include CD28H, VSIG-3 and VSIG-8.
Specifikationer
| VISTA | |
| Polyclonal | |
| Unconjugated | |
| VSIR | |
| 4632428N05Rik; B7H5; B7-H5; C10orf54; C1H10orf54; C28H10orf54; C4H10orf54; chromosome 10 open reading frame 54; DD1alpha; Death Domain1alpha; Dies1; differentiation of ESCs 1; GI24; PD-1H; PDCD1 homolog; PD-H1; platelet receptor Gi24; PP2135; RIKEN cDNA 4632428N05 gene; SISP1; sisp-1; Stress-induced secreted protein-1; UNQ730/PRO1412; V-domain Ig suppressor of T cell activation; VISTA; V-set domain-containing immunoregulatory receptor; V-set immunoregulatory receptor; VSIR; V-type immunoglobulin domain-containing suppressor of T-cell activation | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 64115 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H7M9 | |
| VSIR | |
| Recombinant protein corresponding to Human C10orf54. Recombinant protein control fragment (Product #RP-90094). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering