Learn More
VistaLab™ Technologies Seal Kits
For use with Ovation.
Brand: VistaLab™ Technologies 9057-1003
Additional Details : Weight : 0.10000kg
Description
Contain lubricant, allen wrench, seals and instructionsSpecifications
Standard Seal Kit | |
Ovation™ 2 to 20 μL (Adjustable), 11 to 20 μL (Fixed) Pipette | |
1 Ea. | |
3 in. |
20 μL | |
Lubricant, allen wrench, seals and instructions | |
5 in. | |
1 in. |