Learn More
VistaLab™ Technologies Seal Kits
For use with Ovation.
Brand:  VistaLab™ Technologies 9057-2034
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 11986022
  • 391.56 SEK / 1 set
Estimated Shipment: 07-05-2024
to see stock.
Add to basket
Description
Description
Contain lubricant, allen wrench, seals and instructions
Specifications
Specifications
Viton Seal Kit
Ovation™ 10 to 100 μL (Adjustable), 21 to 100 μL (Fixed) Pipette
1 Ea.
3 in.
10 to 100 μL
Lubricant, allen wrench, seals and instructions
5 in.
1 in.