Läs mer
Vitamin D2 Sulfate Sodium Salt, TRC
Produktkod. 30451587
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30451587 100 mg
1 options
Produktkod. 30451587 Leverantör Toronto Research Chemicals Leverantörsnummer TRCV676030100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Vitamin D2 Sulfate Sodium Salt
CAS 85704-61-6
Molekylformel C28H43NaO4S
Kvantitet 100 mg
Synonym (3β,5Z,7E,22E)-9,10-Secoergosta-5,7,10(19),22-tetraen-3-ol Hydrogen Sulfate, Ergocalciferol Hydrogen Sulfate, Vitamin D2 3β-Sulfate, Vitamin D2 Sulfate, (3beta,5Z,7E,22E)-9,10-Secoergosta-5,7,10(19),22-tetraen-3-ol Hydrogen Sulfate, Vitamin D2 3beta-Sulfate
InChI formel InChI=1S/C28H44O4S.Na/c1-19(2)20(3)9-10-22(5)26-15-16-27-23(8-7-17-28(26,27)6)12-13-24-18-25(14-11-21(24)4)32-33(29,30)31;/h9-10,12-13,19-20,22,25-27H,4,7-8,11,14-18H2,1-3,5-6H3,(H,29,30,31);/q;+1/p-1/b10-9+,23-12+,24-13-;/t20-,22+,25-,26+,27-,28+;/m0./s1
LEDER [H][C@@]1(CC[C@]2([H])[C@@]1(CCC/C2=C\C=C3C[C@H](CCC\3=C)OS(=O)(O[Na])=O)C)[C@@H](/C=C/[C@@H](C(C)C)C)C
IUPAC-namn sodium (S,Z)-3-(2-((1R,3aS,7aR,E)-1-((2R,5R,E)-5,6-dimethylhept-3-en-2-yl)-7a-methyloctahydro-4H-inden-4-ylidene)ethylidene)-4-methylenecyclohexyl sulfate
Molekylvikt (g/mol) 498.69
Formel vikt 498.28