Läs mer
Vitamin D3 Octanoate (90%), TRC
Produktkod. 30454777
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30454777 10 mg
1 options
Produktkod. 30454777 Leverantör Toronto Research Chemicals Leverantörsnummer TRCV67610510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Vitamin D3 Octanoate
CAS 927822-16-0
Molekylformel C35 H58 O2
Kvantitet 10 mg
Synonym Octanoic acid','(1S,3Z)-3-[(2E)-2-[(1R,3aS,7aR)-1-[(1R)-1,5-dimethylhexyl]octahydro-7a-methyl-4H-inden-4-ylidene]ethylidene]-4-methylenecyclohexyl ester (ACI), (1S,3Z)-3-[(2E)-2-[(1R,3aS,7aR)-1-[(1R)-1,5-Dimethylhexyl]octahydro-7a-methyl-4H-inden-4-ylidene]ethylidene]-4-methylenecyclohexyl octanoate (ACI), Octanoic acid (3β,5Z,7E)-9,10-secocholesta-5,7,10(19)-trien-3-yl ester, Cholecalciferol octanoate, Vitamin D3 octanoate
InChI formel InChI=1S/C35H58O2/c1-7-8-9-10-11-17-34(36)37-31-21-18-27(4)30(25-31)20-19-29-16-13-24-35(6)32(22-23-33(29)35)28(5)15-12-14-26(2)3/h19-20,26,28,31-33H,4,7-18,21-25H2,1-3,5-6H3/b29-19+,30-20-/t28-,31+,32-,33+,35-/m1/s1
LEDER CCCCCCCC(=O)O[C@H]1CCC(=C)\C(=C/C=C/2\CCC[C@]3(C)[C@H](CC[C@@H]23)[C@H](C)CCCC(C)C)\C1
Molekylvikt (g/mol) 510.83
Formel vikt 510.4437