Läs mer
Vitamin D3 Sulfate Sodium Salt (Technical Grade), TRC
Produktkod. 30453597
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30453597 1 mg
1 options
Produktkod. 30453597 Leverantör Toronto Research Chemicals Leverantörsnummer TRCV6771601MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Vitamin D3 sulfate sodium salt
CAS 78392-27-5
Molekylformel C27 H43 O4 S . Na
Kvantitet 1 mg
Synonym (1S,3Z)-3-[(2E)-2-[(1R,3aS,7aR)-1-[(1R)-1,5-Dimethylhexyl]octahydro-7a-methyl-4H-inden-4-ylidene]ethylidene]-4-methylenecyclohexanol Sodium Salt, (3β,5Z,7E)-9,10-Secocholesta-5,7,10(19)-trien-3-ol Hydrogen Sulfate Sodium Salt, Cholecalciferol Sulfate Sodium Salt, Vitamin D3 3β-Sulfate Sodium Salt
InChI formel InChI=1S/C27H44O4S.Na/c1-19(2)8-6-9-21(4)25-15-16-26-22(10-7-17-27(25,26)5)12-13-23-18-24(14-11-20(23)3)31-32(28,29)30;/h12-13,19,21,24-26H,3,6-11,14-18H2,1-2,4-5H3,(H,28,29,30);/q;+1/p-1/b22-12+,23-13-;/t21-,24+,25-,26+,27-;/m1./s1
LEDER [Na+].CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2\C(=C\C=C/3\C[C@H](CCC3=C)OS(=O)(=O)[O-])\CCC[C@]12C
IUPAC-namn sodium;[(1S,3Z)-3-[(2E)-2-[(1R,3aS,7aR)-7a-methyl-1-[(2R)-6-methylheptan-2-yl]-2,3,3a,5,6,7-hexahydro-1H-inden-4-ylidene]ethylidene]-4-methylidenecyclohexyl] sulfate
Molekylvikt (g/mol) 486.68
Formel vikt 486.278