Learn More
Vitamin K1, MP Biomedicals™
Click to view available options
Quantity:
1 g
5 g
10 g
Specifications
Color | Yellow |
Density | 0.984 |
Quantity | 1 g |
Formula Weight | 450.68 |
Physical Form | Viscous Liquid |
Chemical Name or Material | Vitamin K1 |