Läs mer
Vitamin K2(5), TRC
Produktkod. 30449876
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30449876 50 mg
30451252 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30449876 Leverantör Toronto Research Chemicals Leverantörsnummer TRCV67612050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 957-78-8
Molekylformel C16H16O2
Molekylvikt (g/mol) 240.3
Synonym 2-Methyl-3-(3-methyl-2-buten-1-yl)-1,4-naphthalenedione, 2-Methyl-3-(3-methyl-2-butenyl)-1,4-naphthalenedione, 2-Methyl-3-(3-methyl-2-butenyl)-1,4-naphthoquinone, Menaquinone 1, NSC 123509, Vitamin MK 1
IUPAC-namn 2-methyl-3-(3-methylbut-2-enyl)naphthalene-1,4-dione
LEDER CC(=CCC1=C(C)C(=O)c2ccccc2C1=O)C
Specifikationer
Kemiskt namn eller material Vitamin K2(5)
Kvantitet 50 mg
InChI formel InChI=1S/C16H16O2/c1-10(2)8-9-12-11(3)15(17)13-6-4-5-7-14(13)16(12)18/h4-8H,9H2,1-3H3
Formel vikt 240.115