Läs mer
Vitamin K4, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Acetomenaphthone (Menadiol Diacetate) |
| CAS | 573-20-6 |
| Molekylformel | C15 H14 O4 |
| Kvantitet | 10 g |
| Synonym | 1,4-Naphthalenediol','2-methyl-','1,4-diacetate1,4-Naphthalenediol','2-methyl-','diacetate (7CI,8CI,9CI), Vitamin K4 (6CI), 1,4-Diacetoxy-2-methylnaphthalene, 2-Methyl-1,4-naphthohydroquinone diacetate, 2-Methyl-1,4-naphthoquinol diacetate, Acetomenaphthone, Adaprin, Davitamon-K, Davitamon-K-oral, Kapathrom, Kapilin, Kapilon, Kappaxan, Kativ powder, Kayvite, Menadiol diacetate, NSC 403062, Pafavit, Prokayvit Oral, Vitamin K diacetate, Vitavel K |
| InChI formel | InChI=1S/C15H14O4/c1-9-8-14(18-10(2)16)12-6-4-5-7-13(12)15(9)19-11(3)17/h4-8H,1-3H3 |
| LEDER | CC(=O)Oc1cc(C)c(OC(=O)C)c2ccccc12 |
| IUPAC-namn | (4-acetyloxy-3-methylnaphthalen-1-yl) acetate |
| Molekylvikt (g/mol) | 258.27 |
| Formel vikt | 258.0892 |