missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VKORC1L1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA562881
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: LIINYKRLVY LNEAWKRQLQ PKQD Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
Involved in vitamin K metabolism. Can reduce inactive vitamin K 2,3-epoxide to active vitamin K (in vitro), and may contribute to vitamin K-mediated protection against oxidative stress. Plays a role in vitamin K-dependent gamma-carboxylation of Glu residues in target proteins.Specifications
VKORC1L1 | |
Polyclonal | |
Unconjugated | |
VKORC1L1 | |
2310024K08Rik; vitamin K epoxide reductase complex subunit 1 like 1; vitamin K epoxide reductase complex subunit 1-like protein 1; vitamin K epoxide reductase complex, subunit 1-like 1; VKORC1L1; VKORC1-like protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
154807 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N0U8 | |
VKORC1L1 | |
Recombinant protein corresponding to Human VKORC1L1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only