missing translation for 'onlineSavingsMsg'
Learn More

VMO1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 21577-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16891175

  • 1656.73 SEK / 20µL
Estimated Shipment: 31-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

VMO1 (Vitelline Membrane Outer Layer 1 Homolog) is a Protein Coding gene.
Specifications

Specifications

VMO1
Polyclonal
Unconjugated
VMO1
ERGA6350, PRO21055, VMO1, vmo-1
Rabbit
Antigen Affinity Chromatography
RUO
284013, 327956, 360553
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Western Blot
0.14 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q5SXG7, Q7Z5L0
VMO1
VMO1 Fusion Protein Ag16120
20 μL
Primary
Rat, Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers