missing translation for 'onlineSavingsMsg'
Learn More

VMO1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 21577-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16881175

  • 3248.87 SEK / 150µL
Estimated Shipment: 30-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

VMO1 (Vitelline Membrane Outer Layer 1 Homolog) is a Protein Coding gene.
Specifications

Specifications

VMO1
0.14 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q5SXG7, Q7Z5L0
ERGA6350, PRO21055, VMO1, vmo-1
Rabbit
IgG
150 μL
-20°C
Primary
284013, 327956, 360553
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
VMO1
Liquid
VMO1
VMO1 Fusion Protein Ag16120
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
SDS
Documents
Certificates
Special Offers

Special Offers