Läs mer
Voglibose, TRC
Produktkod. 30449083
Change view
Kvantitet:
10 mg
100 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30449083 1 g
30450485 10 mg
30449728 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30449083 Leverantör Toronto Research Chemicals Leverantörsnummer TRCV7500001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 83480-29-9
Molekylformel C10 H21 N O7
Molekylvikt (g/mol) 267.28
Synonym 3,4-Dideoxy-4-[[2-hydroxy-1-(hydroxymethyl)ethyl]amino]-2-C-(hydroxymethyl)-D-epi-inositol, (1S,2S,3R,4S,5S)-5-((1,3-Dihydroxypropan-2-yl)amino)-1-(hydroxymethyl)cyclohexane-1,2,3,4-tetraol, (1S,2S,3R,4S,5S)-5-[(1,3-Dihydroxypropan-2-yl)amino]-1-(hydroxymethyl)cyclohexane-1,2,3,4-tetraol, A 71100, AO 128, Basen, Glustat, N-(1,3-Dihydroxy-2-propyl)valiolamine, Voglibose, Voglistat
IUPAC-namn (1S,2S,3R,4S,5S)-5-(1,3-dihydroxypropan-2-ylamino)-1-(hydroxymethyl)cyclohexane-1,2,3,4-tetrol
LEDER OCC(CO)N[C@H]1C[C@](O)(CO)[C@@H](O)[C@H](O)[C@H]1O
Specifikationer
Kemiskt namn eller material Voglibose
Anteckningar om renhetsgrad HPLC
Kvantitet 1 g
InChI formel InChI=1S/C10H21NO7/c12-2-5(3-13)11-6-1-10(18,4-14)9(17)8(16)7(6)15/h5-9,11-18H,1-4H2/t6-,7-,8+,9-,10-/m0/s1
Formel vikt 267.1318
Procent renhet >95%