missing translation for 'onlineSavingsMsg'
Learn More

Karl Hecht™ Volumetric Flask, Conformity Certified

Simply and accurately measure required sample volumes. Karl Hecht™ Volumetric Flask, Conformity Certified is thick-walled, clearly marked and labeled, and has a solid finish.

Brand:  Karl Hecht™ 42097020

Additional Details : Weight : 0.20000kg

 View more versions of this product

Product Code. 15566529

  • 118.72 SEK / Each
Estimated Shipment: 28-06-2024
to see stock.
Add to basket

Description

Description

  • Borosilicate glass 3.3.
  • Fused blue graduation.
  • Standard ground without stopper.
  • Conformity certified.
    • Individual test- or lot-control certificate available from manufacturer with surcharge.
Specifications

Specifications

DIN EN ISO 1042
Borosilicate Glass (3.3)
Volumetric Flask
Yes
20 mL
Standard
0.04 mL
NS 10/19
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers