missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Von Willebrand Factor Antibody (VWF/1767), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54497AF594
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Von Willebrand Factor Monoclonal specifically detects Von Willebrand Factor in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifikationer
| Von Willebrand Factor | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| VWF | |
| Recombinant fragment (around aa1815-1939) of human Von Willebrand Factor protein (exact sequence is proprietary) (Uniprot: P04275) | |
| 0.1 mL | |
| 7450.0 | |
| Human | |
| IgG2b κ |
| Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| VWF/1767 | |
| Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| coagulation factor VIII VWF, F8, F8VWF, von Willebrand factor, VWD, vWF | |
| Mouse | |
| Protein A or G purified | |
| Primary | |
| von Willebrand Factor (vWF) is a multimeric glycoprotein that is found in endothelial cells, plasma and platelets. It acts as a carrier protein for Factor VIII and promotes platelet adhesion and aggregation. vWF undergoes a variety of posttranslational modifications that influence the affinity and availability for Factor VIII, including cleavage of the propeptide and formation of N-terminal disulfide bonds. This antibody helps to establish the endothelial nature of some lesions of disputed histogenesis, e.g. Kaposis sarcoma and cardiac myxoma. It is widely used for differentiating vascular lesions from those of other tissue differentiation within a panel of other vascular markers although not all tumors of endothelial differentiation contain this antigen. | |
| Store at 4°C in the dark. |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering