Learn More
Scientific Industries SI™ Vortex-Genie™ 2
Mixes gently, vigorously, and anywhere in between. Scientific Industries SI™ Vortex-Genie™ 2 speeds range from 600 to 3200rpm.
Brand:  Scientific Industries SI™ SI-0276
Additional Details : Weight : 4.58000kg
 View more versions of this product
Product Code. 15567335
  • 4012.80 SEK / Each
Estimated Shipment
05-01-2021
Add to basket
Description
Description
  • Variable speed for precise mixing
  • Broad range of attachments available for most tubes, plates and other containers
  • Hands-Free or Touch On control
  • May be used in cold rooms and incubators
Specifications
Specifications
165mm
SI-0276
230V
165mm
Swiss
Vortex mixer
230V/60Hz
122mm
600 to 3200rpm
60Hz
Hands-free, Touch on
Pop-off cup, 76mm platform
4kg