Learn More
Scientific Industries SI™ Vortex-Genie™ 2T
Includes an integrated timer. Scientific Industries SI™ Vortex-Genie™ 2T mixes gently or vigorously with a set time or continuously.
Brand:  Scientific Industries SI™ SI-T276
Additional Details : Weight : 4.58000kg
 View more versions of this product
Product Code. 15557365
  • 4878.72 SEK / Each
Estimated Shipment
15-01-2021
Add to basket
Description
Description
  • Variable speed for precise mixing from gentle to vigorous
  • Broad range of attachments for most tubes, plates and other containers
  • Hands-Free or Touch On control
  • May be used in cold rooms and incubators
  • Timer function:
    • Touch On: 1 to 60 sec.
    • Hands-Free: 1 to 60 min. or continuous
  • Unit automatically stops at the end of user-programmed time
Specifications
Specifications
165mm
SI-T276
230V
165mm
76mm platform, pop-off cup
230V/50Hz
122mm
600 to 2700rpm
50Hz
Swiss
4kg