Läs mer
Voxelotor, MedChemExpress
Produktkod. 16487506
Encompass_Preferred
Change view
Kvantitet:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16487506 100 mg
16473597 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16487506 Leverantör Medchem Express Leverantörsnummer HY18681
MedChemExpress Voxelotor (GBT 440) is a potent inhibitor of haemoglobin S (HbS) polymerization. Voxelotor has the potential for sickle cell disease (SCD) treatment.
Voxelotor (GBT 440) is a potent inhibitor of haemoglobin S (HbS) polymerization. Voxelotor has the potential for sickle cell disease (SCD) treatment.
Specifikationer
Kemiskt namn eller material Voxelotor
CAS 1446321-46-5
Färg Light Yellow
Anteckningar om renhetsgrad Research
Molekylformel C19H19N3O3
Kvantitet 100 mg
Synonym GBT 440
Löslighetsinformation DMSO : 100 mg/mL (296.41 mM; Need ultrasonic)
LEDER OC1=CC=CC(OCC2=C(C3=CC=NN3C(C)C)N=CC=C2)=C1C=O
Molekylvikt (g/mol) 337.37
Formel vikt 337.37
Procent renhet 98.23%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) COVID-19-immunoregulation