missing translation for 'onlineSavingsMsg'
Learn More

VPS16 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 17776-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16827754

  • 4721.67 SEK / 150µL
Estimated Shipment: 24-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Vesicle mediated protein sorting plays an important role in segregation of intracellular molecules into distinct organelles. In yeast, Vps proteins are involved in the trafficking of endocytic and biosynthetic proteins to the vacuole, which functionally resembles the lysosome of higher organisms. Human VPS16 is the homolog of yeast class C Vps16 protein. VPS16 may play a role in vesicle-mediated protein trafficking to lysosomal compartments and in membrane docking/fusion reactions of late endosomes/lysosomes.
Specifications

Specifications

VPS16
Polyclonal
Unconjugated
VPS16
hVPS16, VPS16
Rabbit
Antigen Affinity Chromatography
RUO
296159, 64601, 80743
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q920Q4, Q9H269
VPS16
VPS16 Fusion Protein Ag12202
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers