missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ VPS33A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100363
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Antibody detects endogenous levels of total VPS33A.
Vesicle mediated protein sorting plays an important role in segregation of intracellular molecules into distinct organelles. Genetic studies in yeast have identified more than 40 vacuolar protein sorting (VPS) genes involved in vesicle transport to vacuoles. This gene is a member of the Sec-1 domain family, and it encodes a protein similar to the yeast class C Vps33 protein. The mammalian class C VPS proteins are predominantly associated with late endosomes/lysosomes, and like their yeast counterparts, may mediate vesicle trafficking steps in the endosome/lysosome pathway.
Specifikationer
| VPS33A | |
| Polyclonal | |
| Unconjugated | |
| Vps33a | |
| 3830421M04Rik; AI503300; AW048546; AW554476; bf; Buff; hVPS33A; im:7136893; r-Vps33a; similar to vacuolar protein sorting 33A; vacuolar protein sorting 33 homolog A; vacuolar protein sorting 33 homolog A (S. cerevisiae); vacuolar protein sorting 33A; vacuolar protein sorting 33A (yeast); vacuolar protein sorting protein 33a; vacuolar protein sorting-associated protein 33A; vacuolar sorting protein 33a; VPS33A; VPS33A core subunit of CORVET and HOPS complexes; VPS33A CORVET/HOPS core subunit; VPS33A, CORVET/HOPS core subunit; wu:fc08g01; wu:fc26c10 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 65081, 65082, 77573 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q63615, Q96AX1, Q9D2N9 | |
| Vps33a | |
| A synthesized peptide derived from human VPS33A(Accession Q96AX1), corresponding to amino acid residues L499-F549. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering