Läs mer
Invitrogen™ VPS33A Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA531456
Beskrivning
Recommended positive controls: 293T, HeLa, HepG2. Predicted reactivity: Mouse (97%), Rat (96%), Zebrafish (83%), Xenopus laevis (86%), Pig (97%), Rhesus Monkey (100%), Chimpanzee (100%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Vesicle mediated protein sorting plays an important role in segregation of intracellular molecules into distinct organelles. Genetic studies in yeast have identified more than 40 vacuolar protein sorting (VPS) genes involved in vesicle transport to vacuoles. This gene is a member of the Sec-1 domain family, and it encodes a protein similar to the yeast class C Vps33 protein. The mammalian class C VPS proteins are predominantly associated with late endosomes/lysosomes, and like their yeast counterparts, may mediate vesicle trafficking steps in the endosome/lysosome pathway.
Specifikationer
| VPS33A | |
| Polyclonal | |
| Unconjugated | |
| Vps33a | |
| 3830421M04Rik; AI503300; AW048546; AW554476; bf; Buff; hVPS33A; im:7136893; r-Vps33a; similar to vacuolar protein sorting 33A; vacuolar protein sorting 33 homolog A; vacuolar protein sorting 33 homolog A (S. cerevisiae); vacuolar protein sorting 33A; vacuolar protein sorting 33A (yeast); vacuolar protein sorting protein 33a; vacuolar protein sorting-associated protein 33A; vacuolar sorting protein 33a; VPS33A; VPS33A core subunit of CORVET and HOPS complexes; VPS33A CORVET/HOPS core subunit; VPS33A, CORVET/HOPS core subunit; wu:fc08g01; wu:fc26c10 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 65082, 77573 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q96AX1, Q9D2N9 | |
| Vps33a | |
| Recombinant protein encompassing a sequence within the C-terminus region of human VPS33A. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.