missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VPS36 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14262-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
VPS36, VPS22, and VPS25 (Slagsvold et al., 2005 ; Kamura et al., 2001 ).Specifications
VPS36 | |
Polyclonal | |
Unconjugated | |
VPS36 | |
C13orf9, CGI 145, EAP45, ESCRT II complex subunit VPS36, VPS36 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
51028 | |
-20°C | |
Liquid |
Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q86VN1 | |
VPS36 | |
VPS36 Fusion Protein Ag5549 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |