missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VPS37B Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | VPS37B |
---|---|
Concentration | 0.23 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16894914
|
Proteintech
15653-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16884914
|
Proteintech
15653-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Component of the ESCRT-I complex, a regulator of vesicular trafficking process. Required for the sorting of endocytic ubiquitinated cargos into multivesicular bodies. May be involved in cell growth and differentiation.Specifications
VPS37B | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q9H9H4 | |
79720 | |
VPS37B Fusion Protein Ag8256 | |
Primary | |
-20°C | |
VPS37B |
0.23 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
ESCRT I complex subunit VPS37B, hVps37B, VPS37B | |
VPS37B | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |