Läs mer
VRT 043198, TRC
Produktkod. 30455871
Change view
Kvantitet:
1 mg
2.5 mg
5 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30455871 1 mg -
30455287 2.5 mg 2.5mg
30449121 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30455871 Leverantör Toronto Research Chemicals Leverantörsnummer TRCV3750251MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 244133-31-1
Molekylformel C22H29ClN4O6
Molekylvikt (g/mol) 480.18
Synonym VRT 043198, N-(4-Amino-3-chlorobenzoyl)-3-methyl-L-valyl-N-[(1S)-2-carboxy-1-formylethyl]-L-prolinamide
IUPAC-namn (3S)-3-[[(2S)-1-[(2S)-2-[(4-amino-3-chlorobenzoyl)amino]-3,3-dimethylbutanoyl]pyrrolidine-2-carbonyl]amino]-4-oxobutanoic acid
LEDER CC(C)(C)[C@H](NC(=O)c1ccc(N)c(Cl)c1)C(=O)N2CCC[C@H]2C(=O)N[C@@H](CC(=O)O)C=O
Specifikationer
Kemiskt namn eller material N-(4-Amino-3-chlorobenzoyl)-3-methyl-L-valyl-N-[(1S)-2-carboxy-1-formylethyl]-L-prolinamide
Kvantitet 1 mg
InChI formel InChI=1S/C22H29ClN4O6/c1-22(2,3)18(26-19(31)12-6-7-15(24)14(23)9-12)21(33)27-8-4-5-16(27)20(32)25-13(11-28)10-17(29)30/h6-7,9,11,13,16,18H,4-5,8,10,24H2,1-3H3,(H,25,32)(H,26,31)(H,29,30)/t13-,16-,18+/m0/s1
Formel vikt 480.1776