Learn More
IKA™ VSS 1 Vacuum Safety Set
Product Code. 15766678 Shop All IKA Products
Change view
Click to view available options
For Use With (Equipment):
RV Rotary Evaporators, MVP 10 Basic Pump
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
15766678 RV Rotary Evaporators, MVP 10 Basic Pump Each
1 options
Product Code. 15766678 Supplier IKA™ Supplier No. 0020005194
Beräknad leverans: 03-02-2027
Condensate trap connected ahead of the vacuum pup to prevent vapors from being released into the air.
  • Condensate trap system.
  • Prevents vapors from being released into the air.
  • Suitable for use with RV rotary evaporators and the MVP 10 basic pump:
    • RV 3 V
    • RV 3 V-C
    • RV 3 Flex
    • RV 8 V
    • RV 8 V-C
    • RV 8 Flex
    • RV 10 Auto V
    • RV 10 Auto V-C
    • RV 10 Auto Flex
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) RV Rotary Evaporators, MVP 10 Basic Pump