Läs mer
Tocris Bioscience™ VU 0071063
Beskrivning
VU 0071063 is a selective activator of SUR1 Kir6.2 channels and pancreatic β-cell KATP channels (EC50 = 7 μM and 0.3-10.3 μM, respectively). VU 0071063 inhibits insulin secretion in pancreatic β-cells by inducing hyperpolarization of β-cell membranes and reduces voltage-dependent calcium channel activation and Ca2+ influx. VU 0071063 is active in vivo and brain penetrant.
Specifikationer
Specifikationer
| Kemiskt namn eller material | 7-[[4-(1,1-Dimethylethyl)phenyl]methyl]-3,7-dihydro-1,3-dimethyl-1 H-purine-2,6-dione |
| CAS | 333415-38-6 |
| Kvantitet | 10 mg |
| Mål | Inward Rectifier Potassium Channel Activators |
| Molekylformel | C18H22N4O2 |
| Renhet | 0.98 |