Learn More
VU 0365114, MedChemExpress
Product Code. 16486847
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16486847 5 mg 5mg
16485106 100 mg 100mg
16430497 50 mg 50mg
16414148 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16486847 Supplier Medchem Express Supplier No. HY107651
 Restricted Item
Beräknad leverans: 30-11-2026
MedChemExpress VU 0365114 is a mAChR M5 positive allosteric modulator, with an EC50 of 2.7 μM.
VU 0365114 is a mAChR M5 positive allosteric modulator, with an EC50 of 2.7 μM.
Specifications
Chemical Name or Material VU 0365114
CAS 1208222-39-2
Color Orange
Purity Grade Notes Research
Molecular Formula C22H14F3NO3
Quantity 5 mg
Solubility Information DMSO : ≥ 77.5 mg/mL (195.04 mM)
SMILES O=C1N(C2=CC=C(OC(F)(F)F)C=C2C1=O)CC3=CC=C(C4=CC=CC=C4)C=C3
Molecular Weight (g/mol) 397.35
Formula Weight 397.35
Percent Purity 99.41%
Grade Research
Physical Form Solid