Läs mer
Tocris Bioscience™ VU 0469650 Hydrochloride
Potent and selective negative allosteric modulator of mGlu1
Brand: Tocris Bioscience™ 5379/50
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
VU 0469650 hydrochloride is a potent and selective negative allosteric modulator of mGlu1 (IC50 = 99 nM). Exhibits >100-fold selectivity for mGlu1 over mGlu2-8 and 68 other GPCRs, ion channels, kinases and transporters. Brain penetrant.Specifikationer
| VU 0469650 hydrochloride | |
| 3-[(3R)-3-Methyl-4-(tricyclo[3.3.1.13,7]dec-1-ylcarbonyl)-1-piperazinyl]-2-pyridinecarbonitrile hydrochloride | |
| Soluble to 100mM in DMSO and to 50mM in ethanol | |
| Bioactive Small Molecule |
| 1443748-47-7 | |
| 50 mg | |
| Solid |
Produkttitel
Research Use Only. Not For Human or Veterinary Use