missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VWCE Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA550896
Additional Details : Weight : 0.01000kg
Description
The antibody detects endogenous levels of total VWCE protein.
May be a regulatory element in the beta-catenin signaling pathway and a target for chemoprevention of hapatocellular carcinoma.Specifications
VWCE | |
Polyclonal | |
Unconjugated | |
VWCE | |
1300015B04Rik; HBV X protein up-regulated gene 11 protein; HBxAg up-regulated gene 11 protein; RGD1309910; URG11; von Willebrand factor C and EGF domain-containing protein; von Willebrand factor C and EGF domains; VWC1; Vwce | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
220001 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1.8 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q96DN2 | |
VWCE | |
Synthetic peptide of human von Willebrand factor C and EGF domains | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |