Läs mer
VX-702, MedChemExpress
Produktkod. 16420287
Encompass_Preferred
Change view
Kvantitet:
10 mg
50 mg
100 mg
200 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16420287 50 mg
16414956 100 mg
16413928 10 mg
16410986 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16420287 Leverantör Medchem Express Leverantörsnummer HY10401
MedChemExpress VX-702 is a highly selective inhibitor of p38α MAPK, 14-fold higher potency against the p38α versus p38β.
VX-702 is a highly selective inhibitor of p38α MAPK, 14-fold higher potency against the p38α versus p38β.
Specifikationer
Kemiskt namn eller material VX-702
CAS 745833-23-2
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C19H12F4N4O2
Kvantitet 50 mg
Löslighetsinformation DMSO : ≥ 42 mg/mL (103.88 mM)
LEDER O=C(N)C1=CC=C(N=C1C2=CC=C(C=C2F)F)N(C3=C(C=CC=C3F)F)C(N)=O
Molekylvikt (g/mol) 404.32
Formel vikt 404.32
Procent renhet 95.0%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) COVID-19-immunoregulation