Läs mer
VX-765, TRC
Produktkod. 30450147
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30450147 50 mg
30456799 5 mg
30453278 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30450147 Leverantör Toronto Research Chemicals Leverantörsnummer TRCV90060050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 273404-37-8
Molekylformel C24 H33 Cl N4 O6
Molekylvikt (g/mol) 509
Synonym L-Prolinamide, N-(4-amino-3-chlorobenzoyl)-3-methyl-L-valyl-N-[(2R,3S)-2-ethoxytetrahydro-5-oxo-3-furanyl]-, N-(4-Amino-3-chlorobenzoyl)-3-methyl-L-valyl-N-[(2R,3S)-2-ethoxytetrahydro-5-oxo-3-furanyl]-L-prolinamide, Belnacasan, VX 765
IUPAC-namn (2S)-1-[(2S)-2-[(4-amino-3-chlorobenzoyl)amino]-3,3-dimethylbutanoyl]-N-[(2R,3S)-2-ethoxy-5-oxooxolan-3-yl]pyrrolidine-2-carboxamide
LEDER CCO[C@@H]1OC(=O)C[C@@H]1NC(=O)[C@@H]2CCCN2C(=O)[C@@H](NC(=O)c3ccc(N)c(Cl)c3)C(C)(C)C
Specifikationer
Kemiskt namn eller material Belnacasan
Kvantitet 50 mg
InChI formel InChI=1S/C24H33ClN4O6/c1-5-34-23-16(12-18(30)35-23)27-21(32)17-7-6-10-29(17)22(33)19(24(2,3)4)28-20(31)13-8-9-15(26)14(25)11-13/h8-9,11,16-17,19,23H,5-7,10,12,26H2,1-4H3,(H,27,32)(H,28,31)/t16-,17-,19+,23+/m0/s1
Formel vikt 508.2089