missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WARS2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13944-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Aminoacyl-tRNA synthetases catalyze the aminoacylation of tRNA by their cognate amino acid. Because of their central role in linking amino acids with nucleotide triplets contained in tRNAs, aminoacyl-tRNA synthetases are thought to be among the first proteins that appeared in evolution. Two forms of tryptophanyl-tRNA synthetase exist, a cytoplasmic form, named WARS, and a mitochondrial form, named WARS2. This gene encodes the mitochondrial tryptophanyl-tRNA synthetase. Two alternative transcripts encoding different isoforms have been described.Specifications
WARS2 | |
Polyclonal | |
Unconjugated | |
WARS2 | |
(Mt)TrpRS, TrpRS, Tryptophan tRNA ligase, WARS2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10352, 690654, 70560 | |
-20°C | |
Liquid |
Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9CYK1, Q9UGM6 | |
Wars2 | |
WARS2 Fusion Protein Ag5024 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |