Learn More
Macherey-Nagel™ Wash Buffer RA3 Concentrate
Product Code. 15849077
Click to view available options
Quantity:
25 mL
80 mL
Chemical Name or Material:
RA3 Concentrate
RAW2 Buffer
Unit Size:
25mL
80mL
Product Code. 15849077
Brand: Macherey-Nagel™ 740364.80
Supplemental wash buffer used to wash the silica membrane in NucleoSpin™ RNA kits when purifying total RNA.
  • For use with NucleoSpin RNA kits, including: RNA, RNA Plant, RNA / Protein and RNA Clean-up kits
  • Wash buffers are used to wash the silica membrane filter after binding, prior to eluting the RNA
  • Supplemental buffers can be purchased separately
Specifications
Chemical Name or Material RAW2 Buffer
CAS 50-01-1
Color Colorless
For Use With (Application) Purifying total RNA
For Use With (Equipment) NucleoSpin™ RNA kits
pH 6.5 to 7.5
Physical Form Liquid
Quantity 80 mL
Formulation Guanidine hydrochloride, Ethanol, Water
Odor Alcoholic
compliance-icons compliance-icons
Product Identifier
  • Buffer RAW2 (80 mL)
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
Supplemental information
  • MIXTURE LIST-Contains : ethanol, guanidine hydrochloride