missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Washing Buffer I for GeneJET Genomic and Plant Genomic DNA Purification Kits (concentrated), Thermo Scientific™
Handla allt Thermo Scientific ProdukterBeskrivning
Thermo Scientific Washing Buffer I (concentrated) is a component of the GeneJET Genomic DNA Purification Kit (K0721/K0722) and the GeneJET Plant Genomic DNA Purification Mini Kit (K0791/K0792) and may be purchased separately. Provided in 40 mL aliquots.
Specifikationer
Specifikationer
| Kemiskt namn eller material | Wash buffer |
| Beskrivning | Washing Buffer I For GeneJET Genomic and Plant Genomic DNA Purification Kit, 40mL, Concentrated, Liquid, Wash Buffer, Used For DNA Extraction |
| För användning med (applikation) | DNA and RNA purification and analysis, DNA extraction, genomic DNA purification, gDNA from animal tissues and cells, gDNA from bacteria, gDNA from plant and fungi, gDNA from buccal cells |
| Fysisk form | Liquid |
| Kvantitet | 40 ml |
| Formulering | Liquid |
For Research Use Only. Not for use in diagnostic procedures.
Produkttitel
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?