Läs mer
Si Analytics™ Water Distillation Units
Produktkod. 10098281 Handla allt SI Analytics Produkter
Change view
Kapacitet (metrisk):
4 l
7 l
Förpackningsstorlek:
Varje
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kapacitet (metrisk) unitSize
10098281 7 l Varje
10098271 7 l Varje
10407742 4 l Varje
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 10098281 Leverantör Si Analytics™ Leverantörsnummer 285502044
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Incurre no costs for regeneration or replacement of resins, or for pretreatment modules that are required to protect the membrane. SI Analytics™ Water Distillation Unit is easy to clean and ensures maximum operating safety.
  • With the water distilling apparatus made of DURAN™ borosilicate glass 3.3 (hydrolytic class 1 glass), starting with normal tap water you can obtain a distillate of maximum purity that is substantially free from dissolved salts and of extremely low germ content.
  • A maximum conductivity of up to approx. 1μS/cm (25°C) is achieved in the distilled water.
  • The immersion heater is made of high-quality stainless steel, so that the presence of heavy-metal ions in the distillate is excluded.
  • The apparatus saves energy and water, because the cooling water that is heated up in the cooling coil is used for feed water.
  • The outflow of distillate is free from carbon dioxide because its temperature is 60°C.
  • Cost-effective in comparison with other water purification systems, such as ion exchangers or reverse osmosis apparatus
  • No costs are incurred for regeneration or replacement of resins, or for pretreatment modules that are required to protect the membrane.
  • The automatic water level control and built-in boil-dry protection ensure maximum operating safety.
  • Cleaning the apparatus is as easy as cleaning a household coffee maker.
  • In the standard version, the compactly built apparatus is supplied including the connecting cable, plug and hoses and is ready for connection.
  • Different versions are available.
TRUSTED_SUSTAINABILITY
Specifikationer
Beskrivning Water Distillation Unit, 7L, 230/23V/A Electrical input power
Elektriska krav 400 V 8 A
Spänning 400 V
Vikt (metrisk) 11 kg
Typ Water Distillation Unit
Kapacitet (metrisk) 7 l
Funktioner Resistance Heating
Höjd (metrisk) 750 mm
Längd (metrisk) 300 mm
Bredd (metrisk) 300 mm