missing translation for 'onlineSavingsMsg'
Learn More

Water, Filtered through a 0.2 micron filter, BAKER ANALYZED™ LC-MS Reagent, J.T. Baker™

Catalog Number p-5383341
Click to view available options
Quantity:
1 L
Packaging:
Narrow Mouth Amber Glass Bottle
This item is not returnable. View return policy
7732-18-5
H2O
18.015
XLYOFNOQVPJJNP-UHFFFAOYSA-N
water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
962
CHEBI:15377
oxidane
O
This item is not returnable. View return policy
7732-18-5
H2O
18.015
XLYOFNOQVPJJNP-UHFFFAOYSA-N
water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
962
CHEBI:15377
oxidane
O

Function tested and optimized for LC/MS applications

    For Use in Liquid Chromatography and Mass Spectrometry; For Laboratory, Research, or Manufacturing Use
Formula Weight 18.02
Grade BAKER ANALYZED™ LC-MS Reagent
Packaging Narrow Mouth Amber Glass Bottle
Quantity 1 L
Specific Gravity 1L = 1.00kg
Chemical Name or Material Water, Filtered through a 0.2 micron filter
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'