missing translation for 'onlineSavingsMsg'
Läs mer

Water, Supelco™

Produktkod. 15853931
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
10 L
30 L
Förpackningsstorlek:
10 L
30 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet Förpackningsstorlek
15853931 30 L 30 L
15843931 10 L 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 15853931 Leverantör Supelco Leverantörsnummer 1153339030
 Begränsad artikel
Beräknad leverans: 28-10-2026
Lägg i varukorgen
Lägg i varukorgen

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

For chromatography (LC-MS Grade) LiChrosolv™

LiChrosolv™ LC-MS grade solvents are specifically formulated to meet the stringent requirements of liquid chromatography-mass spectrometry applications. With exceptional purity, minimal impurities, and low evaporation residue, our solvents ensure reliable and reproducible results in complex analyses. Their excellent UV transparency and stable composition further enhance performance, making them the ideal choice for high-quality analytical outcomes. Designed to support a wide range of analytical workflows, LiChrosolv™ LC-MS grade solvents minimize potential interferences, empowering you to achieve the best possible results in your mass spectrometric analyses. Now Available with Data Matrix Code This high-quality HPLC-grade solvent now features a unique Data Matrix Code on its label, providing digital access to essential product information such as batch number, product number, and expiry date. Unlike standard barcodes, our DMCs feature a unique serial number for each bottle, enabling you to monitor and trace every bottle individually. With standard barcodes, all bottles from the same batch look identical. Our solution gives you true bottle-level traceability for greater control and compliance in the lab. Know more about our data matrix code-labeled HPLC-grade solvents.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 7732-18-5

Specifikationer

Kemiskt namn eller material Water
Smältpunkt 0°C (lit.)
Kokpunkt 100°C
Molekylformel H2O
MDL-nummer MFCD00011332
Molekylvikt (g/mol) 18.02
Kvalitet Not Available
Densitet 1.000 g/mL at 3.98°C
Kvantitet 30 L
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.