Läs mer
Tocris Bioscience™ WAY 181187 oxalate
High affinity and selective 5-HT6 agonist
Brand: Tocris Bioscience™ 5589/50
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
WAY 181187 oxalate is a high affinity and selective 5-HT6 agonist (Ki = 2.2 nM). Displays 60-fold selectivity over other 5-HT and monoamine receptors. Increases firing rate of 5-HT neurons. Modifies sleep and wakefulness profiles following direct injections into the dorsal raphe nucleus, in vivo. Orally bioavailable.Specifikationer
| 1-[(6-Chloroimidazo[2,1-b]thiazol-5-yl)sulfonyl]-1H-indole-3-ethanamine oxalate | |
| 1883548-85-3 | |
| 50 mg |