missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDR81 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24874-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
WD repeat-containing protein 81 (WDR81) is a 1941 amino acid protein, which contains one BEACH domain and five WD repeats. WDR81 is widely expressed in the brain. WDR81 has an association with cerebellar ataxia, mental retardation, and dysequilibrium syndrome 2. WDR81 exists as six isoforms.Specifications
WDR81 | |
Polyclonal | |
Unconjugated | |
WDR81 | |
WD repeat domain 81, WDR81, WDR81 protein | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
124997, 192652 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.41 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q562E7, Q5ND34 | |
Wdr81 | |
WDR81 Fusion Protein Ag21666 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |