missing translation for 'onlineSavingsMsg'
Learn More

Haldenwanger™ Weighing Boats

Brand:  Haldenwanger™ L Als.Sch.160x40x21

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12081872

  • 839.00 SEK / Each

Estimated Shipment: 27-08-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

160 x 40 x 21 mm
160 mm
Weighing boat
21 mm
40 mm
Videos
SDS
Documents
Certificates