Läs mer
Sampling Systems Weighing Boats
Handla allt Sampling Systems Produkter
Kapacitet (metrisk):
25 ml
50 ml
Sterilitet:
Icke-steril
Sterila
Beskrivning
- Made from high quality food grade PS
- Manufactured in a class 8 cleanroom, packed in a class 7 cleanroom
- Sterile and non-sterile options available
- Available from stock
- Fast delivery worldwide
Specifikationer
Specifikationer
| Kapacitet (engelska) | 0.85 oz. |
| Autoklaveringsbar | Not Autoclavable |
| Bottenform | Flat |
| Kapacitet (metrisk) | 25 ml |
| Färg | White |
| Disponibel | Single-use |
| Material | Polystyrene |
| Form | Boat |
| Sterilitet | Sterila |
| Typ | Weighing Boat |