Learn More
Haldenwanger™ Weighing Boats
Brand:  Haldenwanger™ L Als.Sch.120x30x15
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 12071872
  • 638.00 SEK / Each
Estimated Shipment: 20-08-2021
Specifications
120 x 30 x 15 mm
120 mm
Weighing boat
15 mm
30 mm