Learn More
Fisherbrand™ Grade 919 Weighing Paper
Smooth paper and easily crushable.
140.00 SEK - 484.00 SEK
Specifications
For Use With (Application) Weighing of all types of substances such as beet mash, powders and liquids
Weight (Metric) 25g/m2
Material Paper
Product Type Weighing Paper
View More Specs
Products 2
Product Code Brand Weight (Metric) Price Quantity & Availability  
Product Code Brand Weight (Metric) Price Quantity & Availability  
11760106 Fisherbrand™
25g/m2 140.00 SEK
Pack of 1000 View alternative products
Save Up to 
»
Estimated Shipment
09-04-2019
Add to basket
11750106 Fisherbrand™
25g/m2 This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +46 31 352 32 00 for assistance
N/A View alternative products
Save Up to 
»
Description
Description
  • Smooth paper 25g/m2
  • Easily crushable
Specifications
Specifications
Weighing of all types of substances such as beet mash, powders and liquids
25g/m2
Paper
Weighing Paper